Product Details

SNP ID
rs149667237
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:44480196 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCAGGCCCTTCGTGACATGGTGC[G/T]GAAGGAGTACCCCAACCTGAGCACA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TTPAL PubMed Links

Gene Details

Gene
TTPAL
Gene Name
alpha tocopherol transfer protein like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039199.2 333 Missense Mutation CGG,CTG R66L NP_001034288.1
NM_001261839.1 333 Missense Mutation CGG,CTG R66L NP_001248768.1
NM_024331.4 333 Missense Mutation CGG,CTG R66L NP_077307.2
XM_011529043.2 333 Missense Mutation CGG,CTG R66L XP_011527345.1
XM_011529044.2 333 Missense Mutation CGG,CTG R66L XP_011527346.1
XM_011529045.2 333 Missense Mutation CGG,CTG R66L XP_011527347.1
XM_017028063.1 333 Missense Mutation CGG,CTG R66L XP_016883552.1

View Full Product Details