Product Details
- SNP ID
-
rs149942472
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:1443758 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCAGCTGGCTGGGCGGTTATGTTAA[C/G]CGCTGCACGATGACAGCATTGAGCA
- Phenotype
-
MIM: 606610
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
NSFL1C
PubMed Links
Gene Details
- Gene
- NSFL1C
- Gene Name
- NSFL1 cofactor
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001206736.1 |
2529 |
Silent Mutation |
CGC,CGG |
R370R |
NP_001193665.1 |
NM_016143.4 |
2529 |
Silent Mutation |
CGC,CGG |
R368R |
NP_057227.2 |
NM_018839.4 |
2529 |
Silent Mutation |
CGC,CGG |
R337R |
NP_061327.2 |
XM_006723592.2 |
2529 |
Silent Mutation |
CGC,CGG |
R259R |
XP_006723655.1 |
XM_006723593.2 |
2529 |
Silent Mutation |
CGC,CGG |
R259R |
XP_006723656.1 |
XM_006723594.2 |
2529 |
Silent Mutation |
CGC,CGG |
R257R |
XP_006723657.1 |
XM_011529300.1 |
2529 |
Silent Mutation |
CGC,CGG |
R373R |
XP_011527602.1 |
XM_011529301.1 |
2529 |
Silent Mutation |
CGC,CGG |
R371R |
XP_011527603.1 |
XM_011529302.2 |
2529 |
Silent Mutation |
CGC,CGG |
R320R |
XP_011527604.1 |
XM_011529304.1 |
2529 |
Silent Mutation |
CGC,CGG |
R317R |
XP_011527606.1 |
XM_011529305.2 |
2529 |
Intron |
|
|
XP_011527607.1 |
XM_017027956.1 |
2529 |
Silent Mutation |
CGC,CGG |
R318R |
XP_016883445.1 |
XM_017027957.1 |
2529 |
Silent Mutation |
CGC,CGG |
R319R |
XP_016883446.1 |
XM_017027958.1 |
2529 |
Silent Mutation |
CGC,CGG |
R283R |
XP_016883447.1 |
View Full Product Details