Product Details

SNP ID
rs150827336
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:38815334 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTAATTTATCTGAGCACATGCCAA[C/T]ACACAGATATACCTGGATCTTTAAG
Phenotype
MIM: 613275
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PPP1R16B PubMed Links

Gene Details

Gene
PPP1R16B
Gene Name
protein phosphatase 1 regulatory subunit 16B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172735.2 Intron NP_001166206.1
NM_015568.3 Intron NP_056383.1
XM_011528768.2 Intron XP_011527070.1
XM_011528769.2 Intron XP_011527071.1
XM_017027785.1 Intron XP_016883274.1

View Full Product Details