Product Details

SNP ID
rs151114691
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:23444007 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATGCAAGAGGAATGACACGAGCA[A/G]TTCTTCCTGCCCCCTGCAAAGCAAG
Phenotype
MIM: 609731
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CST11 PubMed Links

Gene Details

Gene
CST11
Gene Name
cystatin 11
There are no transcripts associated with this gene.

Gene
CSTL1
Gene Name
cystatin like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138283.1 539 Missense Mutation AAT,AGT N98S NP_612140.1

View Full Product Details