Product Details

SNP ID
rs151318996
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:62389482 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTGTCCTGCACCACTGCTGGGGT[C/T]GGGTGGAGTTGCTGGTGGAGTTCTG
Phenotype
MIM: 180477
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CABLES2 PubMed Links

Gene Details

Gene
CABLES2
Gene Name
Cdk5 and Abl enzyme substrate 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031215.2 2933 UTR 3 NP_112492.2
Gene
RPS21
Gene Name
ribosomal protein S21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024.3 2933 Intron NP_001015.1

View Full Product Details