Product Details

SNP ID
rs138078875
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:44806044 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACTTGGAAGTAGCTCTTATACAC[A/G]ACAACATAACTGCAAATTCAGCAAT
Phenotype
MIM: 602231 MIM: 603124
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC01424 PubMed Links

Gene Details

Gene
LINC01424
Gene Name
long intergenic non-protein coding RNA 1424
There are no transcripts associated with this gene.

Gene
SUMO3
Gene Name
small ubiquitin-like modifier 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286416.1 1380 UTR 3 NP_001273345.1
NM_006936.2 1380 UTR 3 NP_008867.2
Gene
UBE2G2
Gene Name
ubiquitin conjugating enzyme E2 G2
There are no transcripts associated with this gene.

View Full Product Details