Product Details

SNP ID
rs139070825
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:21009141 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTATTCCTAGAACATGGTAGAAGCC[C/T]GGCAAATATGTGCTGAATGCATATT
Phenotype
MIM: 602040
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NCAM2 PubMed Links

Gene Details

Gene
NCAM2
Gene Name
neural cell adhesion molecule 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004540.3 Intron NP_004531.2
XM_011529575.2 Intron XP_011527877.1
XM_011529576.2 Intron XP_011527878.1
XM_011529579.2 Intron XP_011527881.1
XM_011529580.2 Intron XP_011527882.1
XM_011529581.2 Intron XP_011527883.1
XM_011529582.2 Intron XP_011527884.1
XM_011529585.2 Intron XP_011527887.1
XM_017028353.1 Intron XP_016883842.1
XM_017028354.1 Intron XP_016883843.1
XM_017028355.1 Intron XP_016883844.1
XM_017028356.1 Intron XP_016883845.1
XM_017028357.1 Intron XP_016883846.1
XM_017028358.1 Intron XP_016883847.1

View Full Product Details