Product Details

SNP ID
rs139801844
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:30396509 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGCTGTGGCTTCCCTTCCCTGGG[A/C]TATGGCGTTGGATTCTGCCGCCCAA
Phenotype
MIM: 608718
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KRTAP13-1 PubMed Links

Gene Details

Gene
KRTAP13-1
Gene Name
keratin associated protein 13-1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181599.2 436 Silent Mutation GGA,GGC G141G NP_853630.2

View Full Product Details