Product Details

SNP ID
rs139820526
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:44851582 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGGGACTGATGTGCTGGAGCGCTT[C/T]AGTTGTTTTCAAATCTAGCATACGG
Phenotype
MIM: 603784
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PTTG1IP PubMed Links

Gene Details

Gene
PTTG1IP
Gene Name
pituitary tumor-transforming 1 interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286822.1 829 Missense Mutation AAA,GAA K72E NP_001273751.1
NM_004339.3 829 Silent Mutation TAA,TGA *181* NP_004330.1

View Full Product Details