Product Details
- SNP ID
-
rs140855174
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:46138897 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGCTCACCTGTCCTTTTCCTCAGG[G/T]GTGTTCTTGGGGAGCCTCATTGCTT
- Phenotype
-
MIM: 120240
MIM: 606806
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
COL6A2
PubMed Links
Gene Details
- Gene
- COL6A2
- Gene Name
- collagen type VI alpha 2
There are no transcripts associated with this gene.
- Gene
- FTCD
- Gene Name
- formimidoyltransferase cyclodeaminase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001320412.1 |
2374 |
Silent Mutation |
ACA,ACC |
T429T |
NP_001307341.1 |
NM_006657.2 |
2374 |
Silent Mutation |
ACA,ACC |
T429T |
NP_006648.1 |
NM_206965.1 |
2374 |
Silent Mutation |
ACA,ACC |
T429T |
NP_996848.1 |
XM_006723961.3 |
2374 |
Silent Mutation |
ACA,ACC |
T512T |
XP_006724024.2 |
XM_006723962.3 |
2374 |
Silent Mutation |
ACA,ACC |
T512T |
XP_006724025.2 |
XM_011529434.2 |
2374 |
Silent Mutation |
ACA,ACC |
T512T |
XP_011527736.1 |
XM_011529435.2 |
2374 |
Silent Mutation |
ACA,ACC |
T469T |
XP_011527737.1 |
XM_011529436.2 |
2374 |
Silent Mutation |
ACA,ACC |
T512T |
XP_011527738.1 |
XM_011529437.2 |
2374 |
Silent Mutation |
ACA,ACC |
T512T |
XP_011527739.1 |
XM_011529439.2 |
2374 |
Silent Mutation |
ACA,ACC |
T341T |
XP_011527741.1 |
XM_011529440.2 |
2374 |
Intron |
|
|
XP_011527742.1 |
View Full Product Details