Product Details

SNP ID
rs142513484
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:25585733 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGGTAGATGTACATTCCTCTGTTG[C/T]TTTACTTTGATCTTCAGTTACCTGT
Phenotype
MIM: 611845
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00515 PubMed Links

Gene Details

Gene
LINC00515
Gene Name
long intergenic non-protein coding RNA 515
There are no transcripts associated with this gene.

Gene
MRPL39
Gene Name
mitochondrial ribosomal protein L39
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017446.3 1005 Missense Mutation ACA,GCA T331A NP_059142.2
NM_080794.3 1005 UTR 3 NP_542984.2
XM_006724026.3 1005 Intron XP_006724089.1
XM_011529651.2 1005 Missense Mutation ACA,GCA T289A XP_011527953.1

View Full Product Details