Product Details

SNP ID
rs142705847
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:44647250 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGTGCCCCCACCTCCTCCTGCCA[A/G]TCCAGCTGCTGCCGCCCGGCCTCCT
Phenotype
MIM: 612920
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KRTAP10-10 PubMed Links

Gene Details

Gene
KRTAP10-10
Gene Name
keratin associated protein 10-10
There are no transcripts associated with this gene.

Gene
KRTAP10-11
Gene Name
keratin associated protein 10-11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198692.2 837 Silent Mutation CAA,CAG Q264Q NP_941965.2
Gene
KRTAP12-4
Gene Name
keratin associated protein 12-4
There are no transcripts associated with this gene.

Gene
TSPEAR
Gene Name
thrombospondin type laminin G domain and EAR repeats
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001272037.1 837 Intron NP_001258966.1
NM_144991.2 837 Intron NP_659428.2

View Full Product Details