Product Details
- SNP ID
-
rs142788946
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:44287012 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCAGCCAGCCCCGGAAGGGGAGGAA[G/T]CCCCCGGCCGTCCCCAAGGCTTTGG
- Phenotype
-
MIM: 607358
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
AIRE
PubMed Links
Gene Details
- Gene
- AIRE
- Gene Name
- autoimmune regulator
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000383.3 |
511 |
Missense Mutation |
AAG,AAT |
K114N |
NP_000374.1 |
View Full Product Details