Product Details

SNP ID
rs143718199
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:44851462 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTCTCCGGCCGCCTGTCCTGGAA[G/T]GCTGGCAGGTTCACTGGCCAAGGTC
Phenotype
MIM: 603784
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PTTG1IP PubMed Links

Gene Details

Gene
PTTG1IP
Gene Name
pituitary tumor-transforming 1 interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286822.1 949 Missense Mutation NP_001273751.1
NM_004339.3 949 UTR 3 NP_004330.1

View Full Product Details