Product Details

SNP ID
rs146339780
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:46602028 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAATGAGTCTAGCATTATTTGGGA[C/T]GCTTTGGCCTTTGGGACCTTGATGC
Phenotype
MIM: 176990
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
S100B PubMed Links

Gene Details

Gene
S100B
Gene Name
S100 calcium binding protein B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006272.2 Intron NP_006263.1
XM_017028424.1 Intron XP_016883913.1

View Full Product Details