Product Details

SNP ID
rs146532887
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:37227805 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTCGCGGGCATAGCCTTCTGCACA[A/C]CCTGCGGGAGGGAGGCCACATCACG
Phenotype
MIM: 605298
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DSCR3 PubMed Links

Gene Details

Gene
DSCR3
Gene Name
DSCR3 arrestin fold containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006052.1 1409 Silent Mutation GGG,GGT G220G NP_006043.1
XM_005260909.4 1409 Silent Mutation GGG,GGT G193G XP_005260966.1
XM_005260911.4 1409 Silent Mutation GGG,GGT G95G XP_005260968.1
XM_011529423.2 1409 Silent Mutation GGG,GGT G172G XP_011527725.1
Gene
DSCR9
Gene Name
Down syndrome critical region 9 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details