Product Details

SNP ID
rs146999256
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:46235283 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCAGGCTGCTCAAATGTCCACCAT[A/G]TCTAGCAGCGCAGAGAGATGGAGCT
Phenotype
MIM: 600909 MIM: 603294
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LSS PubMed Links

Gene Details

Gene
LSS
Gene Name
lanosterol synthase (2,3-oxidosqualene-lanosterol cyclase)
There are no transcripts associated with this gene.

Gene
MCM3AP
Gene Name
minichromosome maintenance complex component 3 associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003906.4 6508 Silent Mutation NP_003897.2
XM_005261203.4 6508 Silent Mutation XP_005261260.1
XM_005261204.4 6508 Silent Mutation XP_005261261.1
XM_005261205.3 6508 Silent Mutation XP_005261262.1
Gene
MCM3AP-AS1
Gene Name
MCM3AP antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details