Product Details

SNP ID
rs147050202
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:14373089 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTGTGAATGTAAATTTGTTATAT[A/C]ATATGGCAGTTACTGAATATTACGT
Phenotype
MIM: 601100
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HSPA13 PubMed Links

Gene Details

Gene
HSPA13
Gene Name
heat shock protein family A (Hsp70) member 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006948.4 2013 UTR 3 NP_008879.3

View Full Product Details