Product Details

SNP ID
rs138513724
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:23181098 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAAGGCCTCCATTCGGCGCCTGGA[G/T]CAGGAGGTGAACCAGGAGCGCTTCC
Phenotype
MIM: 151410 MIM: 605663
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
BCR PubMed Links

Gene Details

Gene
BCR
Gene Name
BCR, RhoGEF and GTPase activating protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004327.3 734 Missense Mutation GAG,GAT E46D NP_004318.3
NM_021574.2 734 Missense Mutation GAG,GAT E46D NP_067585.2
Gene
RSPH14
Gene Name
radial spoke head 14 homolog
There are no transcripts associated with this gene.

View Full Product Details