Product Details

SNP ID
rs139305630
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:43207558 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTCAAAATTTCCTGGTGGTTCTC[C/T]GAGGCGTACAGGCGCCCATCGCGCA
Phenotype
MIM: 611746
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105373051 PubMed Links

Gene Details

Gene
LOC105373051
Gene Name
uncharacterized LOC105373051
There are no transcripts associated with this gene.

Gene
SCUBE1
Gene Name
signal peptide, CUB domain and EGF like domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173050.3 2917 Silent Mutation TCA,TCG S930S NP_766638.2
XM_005261752.1 2917 Silent Mutation TCA,TCG S960S XP_005261809.1
XM_011530389.2 2917 Silent Mutation TCA,TCG S854S XP_011528691.1
XM_011530390.2 2917 Silent Mutation TCA,TCG S854S XP_011528692.1

View Full Product Details