Product Details

SNP ID
rs139719484
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:46686646 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGATCAAGCAGCGATGTCCTAACC[A/G]TAAACGAAGAGGAAGGTGAAATGAA
Phenotype
MIM: 610307 MIM: 613691
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CERK PubMed Links

Gene Details

Gene
CERK
Gene Name
ceramide kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022766.5 2209 UTR 3 NP_073603.2
XM_017028909.1 2209 UTR 3 XP_016884398.1
Gene
GRAMD4
Gene Name
GRAM domain containing 4
There are no transcripts associated with this gene.

View Full Product Details