Product Details

SNP ID
rs140364088
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:16962332 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCCCCTGACCAAAAAATGCCAGG[A/G]CTGAAAAGGTCCATGTCCACTGTGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GAB4 PubMed Links

Gene Details

Gene
GAB4
Gene Name
GRB2 associated binding protein family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037814.1 2234 UTR 3 NP_001032903.1
XM_011546114.2 2234 Intron XP_011544416.1
XM_011546115.1 2234 Intron XP_011544417.1
XM_011546116.2 2234 Intron XP_011544418.1
XM_011546117.1 2234 Intron XP_011544419.1
XM_017028575.1 2234 Intron XP_016884064.1
XM_017028576.1 2234 Intron XP_016884065.1

View Full Product Details