Product Details

SNP ID
rs141048897
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:46687204 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGAGTCGAACCAGCTGGCAGTGG[A/C]CTCTGCAGAGACAAGCGGCCACGTG
Phenotype
MIM: 610307 MIM: 613691
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CERK PubMed Links

Gene Details

Gene
CERK
Gene Name
ceramide kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022766.5 1651 Missense Mutation GGC,GTC G515V NP_073603.2
XM_017028909.1 1651 Missense Mutation GGC,GTC G478V XP_016884398.1
Gene
GRAMD4
Gene Name
GRAM domain containing 4
There are no transcripts associated with this gene.

View Full Product Details