Product Details

SNP ID
rs141418864
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:39101263 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAGTGCCATGCAGAAATTTGCTT[C/T]ATTAACGAGATCAAGTCCATGGGAC
Phenotype
MIM: 610976
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
APOBEC3H PubMed Links

Gene Details

Gene
APOBEC3H
Gene Name
apolipoprotein B mRNA editing enzyme catalytic subunit 3H
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166002.2 324 Silent Mutation TTC,TTT F59F NP_001159474.2
NM_001166003.2 324 Silent Mutation TTC,TTT F59F NP_001159475.2
NM_001166004.2 324 Silent Mutation TTC,TTT F59F NP_001159476.2
NM_181773.4 324 Silent Mutation TTC,TTT F59F NP_861438.3
XM_011529990.2 324 Silent Mutation TTC,TTT F59F XP_011528292.1
XM_011529991.2 324 Silent Mutation TTC,TTT F59F XP_011528293.1
XM_011529992.2 324 Silent Mutation TTC,TTT F59F XP_011528294.1

View Full Product Details