Product Details
- SNP ID
-
rs143360764
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:29215578 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACAAGGTGATGTACAAGATAGTGAC[C/T]GCCCGTGAGTGGAGGTGCTGCCCTG
- Phenotype
-
MIM: 608926
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
EMID1
PubMed Links
Gene Details
- Gene
- EMID1
- Gene Name
- EMI domain containing 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001267895.1 |
389 |
Silent Mutation |
ACC,ACT |
T89T |
NP_001254824.1 |
NM_133455.3 |
389 |
Silent Mutation |
ACC,ACT |
T89T |
NP_597712.2 |
XM_005261328.2 |
389 |
Silent Mutation |
ACC,ACT |
T89T |
XP_005261385.1 |
XM_005261329.2 |
389 |
Silent Mutation |
ACC,ACT |
T89T |
XP_005261386.1 |
XM_011529868.2 |
389 |
Silent Mutation |
ACC,ACT |
T89T |
XP_011528170.1 |
XM_011529869.2 |
389 |
Silent Mutation |
ACC,ACT |
T89T |
XP_011528171.1 |
XM_011529870.2 |
389 |
Silent Mutation |
ACC,ACT |
T89T |
XP_011528172.1 |
XM_011529871.2 |
389 |
Silent Mutation |
ACC,ACT |
T89T |
XP_011528173.1 |
XM_011529872.2 |
389 |
Silent Mutation |
ACC,ACT |
T89T |
XP_011528174.1 |
XM_011529873.2 |
389 |
Silent Mutation |
ACC,ACT |
T89T |
XP_011528175.1 |
XM_011529874.2 |
389 |
Silent Mutation |
ACC,ACT |
T24T |
XP_011528176.1 |
XM_011529875.2 |
389 |
UTR 5 |
|
|
XP_011528177.1 |
XM_011529876.1 |
389 |
Intron |
|
|
XP_011528178.1 |
XM_017028589.1 |
389 |
Silent Mutation |
ACC,ACT |
T24T |
XP_016884078.1 |
View Full Product Details