Product Details

SNP ID
rs143641709
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:37943061 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACAGGGCCGTTTTATTCTGCAAAC[C/T]GAAACTTGGGTCATGTGACCTGATG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C22orf23 PubMed Links

Gene Details

Gene
C22orf23
Gene Name
chromosome 22 open reading frame 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001207062.1 1766 UTR 3 NP_001193991.1
NM_032561.4 1766 UTR 3 NP_115950.3
XM_005261781.1 1766 Intron XP_005261838.1
XM_005261782.3 1766 Intron XP_005261839.1
XM_005261783.2 1766 Intron XP_005261840.1
XM_005261784.2 1766 Intron XP_005261841.1
Gene
MICALL1
Gene Name
MICAL like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033386.3 1766 Intron NP_203744.1
XM_005261791.3 1766 Intron XP_005261848.1
XM_005261792.3 1766 Intron XP_005261849.1
XM_011530471.2 1766 Intron XP_011528773.1
XM_011530472.2 1766 Intron XP_011528774.1
XM_011530473.2 1766 Intron XP_011528775.1
XM_011530474.2 1766 Intron XP_011528776.1
XM_011530475.2 1766 Intron XP_011528777.1
XM_011530476.2 1766 Intron XP_011528778.1
XM_017029026.1 1766 Intron XP_016884515.1

View Full Product Details