Product Details

SNP ID
rs143928740
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:37642609 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAAGGAGCTGGACCCTGATTCCA[C/G]CATGGGGTGAGCACAGACGGGGCCC
Phenotype
MIM: 606004
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GGA1 PubMed Links

Gene Details

Gene
GGA1
Gene Name
golgi associated, gamma adaptin ear containing, ARF binding protein 1
There are no transcripts associated with this gene.

Gene
LOC101927051
Gene Name
uncharacterized LOC101927051
There are no transcripts associated with this gene.

Gene
SH3BP1
Gene Name
SH3 domain binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018957.3 389 Missense Mutation ACC,AGC T93S NP_061830.3

View Full Product Details