Product Details

SNP ID
rs145014744
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:19022825 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACATACAAATCAGGCCCATCTGGA[A/G]AGAAGGGAGTGAAGCATTGCGCTGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DGCR10 PubMed Links

Gene Details

Gene
DGCR10
Gene Name
DiGeorge syndrome critical region gene 10 (non-protein coding)
There are no transcripts associated with this gene.

Gene
DGCR5
Gene Name
DiGeorge syndrome critical region gene 5 (non-protein coding)
There are no transcripts associated with this gene.

Gene
DGCR9
Gene Name
DiGeorge syndrome critical region gene 9 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details