Product Details
- SNP ID
-
rs146070249
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:50092025 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTGACACTAAGCTGAAAACTGTAC[C/G]GGGTGTCGTGACAAGGTACTGCAGC
- Phenotype
-
MIM: 605908
MIM: 605794
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MLC1
PubMed Links
Gene Details
- Gene
- MLC1
- Gene Name
- megalencephalic leukoencephalopathy with subcortical cysts 1
There are no transcripts associated with this gene.
- Gene
- MOV10L1
- Gene Name
- Mov10 RISC complex RNA helicase like 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001164104.1 |
205 |
Missense Mutation |
CCG,CGG |
P41R |
NP_001157576.1 |
NM_001164105.1 |
205 |
Missense Mutation |
CCG,CGG |
P21R |
NP_001157577.1 |
NM_001164106.1 |
205 |
Intron |
|
|
NP_001157578.1 |
NM_018995.2 |
205 |
Missense Mutation |
CCG,CGG |
P41R |
NP_061868.1 |
XM_005261923.3 |
205 |
Missense Mutation |
CCG,CGG |
P21R |
XP_005261980.1 |
XM_011530696.1 |
205 |
Missense Mutation |
CCG,CGG |
P41R |
XP_011528998.1 |
XM_011530697.1 |
205 |
Missense Mutation |
CCG,CGG |
P21R |
XP_011528999.1 |
XM_011530698.1 |
205 |
Missense Mutation |
CCG,CGG |
P41R |
XP_011529000.1 |
XM_011530699.1 |
205 |
UTR 5 |
|
|
XP_011529001.1 |
XM_011530700.2 |
205 |
UTR 5 |
|
|
XP_011529002.1 |
XM_011530701.1 |
205 |
Missense Mutation |
CCG,CGG |
P41R |
XP_011529003.1 |
XM_011530702.1 |
205 |
UTR 5 |
|
|
XP_011529004.1 |
XM_011530703.1 |
205 |
Intron |
|
|
XP_011529005.1 |
XM_011530704.1 |
205 |
Missense Mutation |
CCG,CGG |
P41R |
XP_011529006.1 |
XM_017028833.1 |
205 |
Missense Mutation |
CCG,CGG |
P41R |
XP_016884322.1 |
XM_017028834.1 |
205 |
Missense Mutation |
CCG,CGG |
P41R |
XP_016884323.1 |
XM_017028835.1 |
205 |
Intron |
|
|
XP_016884324.1 |
XM_017028836.1 |
205 |
Intron |
|
|
XP_016884325.1 |
XM_017028837.1 |
205 |
Missense Mutation |
CCG,CGG |
P41R |
XP_016884326.1 |
View Full Product Details