Product Details

SNP ID
rs146133764
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:19877206 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGTGGGATGGATACCCACGGTCC[A/G]CATCACCTGCGCATAGGAAGCCCCA
Phenotype
MIM: 606448
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TXNRD2 PubMed Links

Gene Details

Gene
TXNRD2
Gene Name
thioredoxin reductase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282512.1 1663 Intron NP_001269441.1
NM_006440.4 1663 Missense Mutation NP_006431.2

View Full Product Details