Product Details

SNP ID
rs146374327
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:42168679 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCCCCGCTCCGACTGCTCTGTGC[A/T]GAGGCTGCCTTTCGCGGTCTTGTTC
Phenotype
MIM: 603107
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TCF20 PubMed Links

Gene Details

Gene
TCF20
Gene Name
transcription factor 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005650.3 6178 Missense Mutation AGC,TGC S1953C NP_005641.1
NM_181492.2 6178 Intron NP_852469.1
XM_005261722.3 6178 Missense Mutation AGC,TGC S1953C XP_005261779.1
XM_006724313.3 6178 Missense Mutation AGC,TGC S1953C XP_006724376.1
XM_011530353.2 6178 Missense Mutation AGC,TGC S1942C XP_011528655.1
XM_011530354.2 6178 Intron XP_011528656.1

View Full Product Details