Product Details

SNP ID
rs146500316
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:29058831 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCTGCCAGGGAACCGGTCCCTCT[A/G]AGCACCTTCAGAGATGGCACCCTGA
Phenotype
MIM: 612062
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C22orf31 PubMed Links

Gene Details

Gene
C22orf31
Gene Name
chromosome 22 open reading frame 31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015370.1 706 Nonsense Mutation CAG,TAG Q262* NP_056185.1
XM_005261491.3 706 Nonsense Mutation CAG,TAG Q158* XP_005261548.1
XM_011530096.2 706 Nonsense Mutation CAG,TAG Q221* XP_011528398.1
XM_017028741.1 706 Nonsense Mutation CAG,TAG Q221* XP_016884230.1
Gene
ZNRF3
Gene Name
zinc and ring finger 3
There are no transcripts associated with this gene.

View Full Product Details