Product Details

SNP ID
rs146817251
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:24935525 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCATAATACATGGAGGAGGCTTC[A/G]CACACTTCCTTGATGAATGGGGAGG
Phenotype
MIM: 611417
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SGSM1 PubMed Links

Gene Details

Gene
SGSM1
Gene Name
small G protein signaling modulator 1
There are no transcripts associated with this gene.

Gene
TMEM211
Gene Name
transmembrane protein 211
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001663.1 223 Silent Mutation NP_001001663.1
XM_011530094.2 223 Silent Mutation XP_011528396.1

View Full Product Details