Product Details

SNP ID
rs147483140
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:41810222 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCCCTCAGGTGCTGGAGGACTCC[C/T]GGACAGTGCTCACCGCTGCTGATGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC134 PubMed Links

Gene Details

Gene
CCDC134
Gene Name
coiled-coil domain containing 134
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304797.1 492 Intron NP_001291726.1
NM_024821.3 492 Missense Mutation CGG,TGG R81W NP_079097.1
XM_005261748.3 492 Missense Mutation CGG,TGG R81W XP_005261805.1

View Full Product Details