Product Details
- SNP ID
-
rs147600184
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:38782689 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGACCAGAGGGAAGGTCTGCAGTC[A/G]CTTATAATCAGCCTCATCTTTCTTC
- Phenotype
-
MIM: 610565
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DNAL4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs760482] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DNAL4
- Gene Name
- dynein axonemal light chain 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005740.2 |
271 |
Nonsense Mutation |
CGA,TGA |
R15* |
NP_005731.1 |
View Full Product Details