Product Details
- SNP ID
-
rs147768775
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:37686410 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCACTGCTATGGGCCGCAACAAGAA[C/G]AAGAAGCGAGATGGTGACGACCGGC
- Phenotype
-
MIM: 150570
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
LGALS1
PubMed Links
Gene Details
- Gene
- LGALS1
- Gene Name
- galectin 1
There are no transcripts associated with this gene.
- Gene
- NOL12
- Gene Name
- nucleolar protein 12
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_024313.2 |
74 |
Missense Mutation |
AAC,AAG |
N6K |
NP_077289.1 |
View Full Product Details