Product Details

SNP ID
rs149250882
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:46685716 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCACCATTAAAGCTGCCCTTTCA[C/G]CTGATCTGGGTGGGGCTAAACGAGG
Phenotype
MIM: 610307 MIM: 613691
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CERK PubMed Links

Gene Details

Gene
CERK
Gene Name
ceramide kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022766.5 3139 UTR 3 NP_073603.2
XM_017028909.1 3139 UTR 3 XP_016884398.1
Gene
GRAMD4
Gene Name
GRAM domain containing 4
There are no transcripts associated with this gene.

View Full Product Details