Product Details

SNP ID
rs151214632
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:20709982 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATGAGAGTGACCAGGGAGACGAC[C/T]GCGTCCATGTAGGGCCTGGGGAGAG
Phenotype
MIM: 600286
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PI4KA PubMed Links

Gene Details

Gene
PI4KA
Gene Name
phosphatidylinositol 4-kinase alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_058004.3 6120 Silent Mutation GCA,GCG A2033A NP_477352.3
XM_005261634.1 6120 Silent Mutation GCA,GCG A2030A XP_005261691.1
XM_005261635.1 6120 Silent Mutation GCA,GCG A1785A XP_005261692.1
XM_011530226.1 6120 Intron XP_011528528.1
XM_011530228.2 6120 Intron XP_011528530.1
XM_017028829.1 6120 Silent Mutation GCA,GCG A2039A XP_016884318.1
XM_017028830.1 6120 Silent Mutation GCA,GCG A1400A XP_016884319.1
Gene
TMEM191A
Gene Name
transmembrane protein 191A (pseudogene)
There are no transcripts associated with this gene.

View Full Product Details