Product Details

SNP ID
rs137964402
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:46907902 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATGCATTTTGAAGTAATGGAGCT[A/G]CAATTCTTGTGGCGACATCTCCGCC
Phenotype
MIM: 607788 MIM: 609332
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MCFD2 PubMed Links

Gene Details

Gene
MCFD2
Gene Name
multiple coagulation factor deficiency 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171506.2 355 Intron NP_001164977.1
NM_001171507.2 355 Nonsense Mutation CAG,TAG Q73* NP_001164978.1
NM_001171508.2 355 Nonsense Mutation CAG,TAG Q73* NP_001164979.1
NM_001171509.2 355 Intron NP_001164980.1
NM_001171510.2 355 Intron NP_001164981.1
NM_001171511.2 355 Nonsense Mutation CAG,TAG Q54* NP_001164982.1
NM_139279.5 355 Intron NP_644808.1
Gene
TTC7A
Gene Name
tetratricopeptide repeat domain 7A
There are no transcripts associated with this gene.

View Full Product Details