Product Details
- SNP ID
-
rs138077680
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:219502418 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCAATCCCTCAACTACGGCTGCATC[A/G]TTGAGAATCCACAGACACACGAGGT
- Phenotype
-
MIM: 615495
MIM: 615950
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GMPPA
PubMed Links
Gene Details
- Gene
- GMPPA
- Gene Name
- GDP-mannose pyrophosphorylase A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_013335.3 |
489 |
Missense Mutation |
ATT,GTT |
I156V |
NP_037467.2 |
NM_205847.2 |
489 |
Missense Mutation |
ATT,GTT |
I156V |
NP_995319.1 |
XM_005246483.3 |
489 |
Missense Mutation |
ATT,GTT |
I156V |
XP_005246540.1 |
XM_005246486.3 |
489 |
Missense Mutation |
ATT,GTT |
I156V |
XP_005246543.1 |
XM_011511032.2 |
489 |
Missense Mutation |
ATT,GTT |
I156V |
XP_011509334.1 |
XM_017003906.1 |
489 |
Missense Mutation |
ATT,GTT |
I156V |
XP_016859395.1 |
XM_017003907.1 |
489 |
Missense Mutation |
ATT,GTT |
I156V |
XP_016859396.1 |
XM_017003908.1 |
489 |
Missense Mutation |
ATT,GTT |
I156V |
XP_016859397.1 |
XM_017003909.1 |
489 |
Missense Mutation |
ATT,GTT |
I156V |
XP_016859398.1 |
XM_017003910.1 |
489 |
Missense Mutation |
ATT,GTT |
I156V |
XP_016859399.1 |
XM_017003911.1 |
489 |
Missense Mutation |
ATT,GTT |
I156V |
XP_016859400.1 |
XM_017003912.1 |
489 |
Missense Mutation |
ATT,GTT |
I156V |
XP_016859401.1 |
XM_017003913.1 |
489 |
Missense Mutation |
ATT,GTT |
I156V |
XP_016859402.1 |
XM_017003914.1 |
489 |
Missense Mutation |
ATT,GTT |
I31V |
XP_016859403.1 |
XM_017003915.1 |
489 |
Missense Mutation |
ATT,GTT |
I31V |
XP_016859404.1 |
XM_017003916.1 |
489 |
Missense Mutation |
ATT,GTT |
I31V |
XP_016859405.1 |
XM_017003917.1 |
489 |
Missense Mutation |
ATT,GTT |
I31V |
XP_016859406.1 |
- Gene
- LOC100996693
- Gene Name
- CAVP-target protein-like
There are no transcripts associated with this gene.
- Gene
- SPEG
- Gene Name
- SPEG complex locus
There are no transcripts associated with this gene.
View Full Product Details