Product Details

SNP ID
rs138509553
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:201626095 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATAATCCAACCAGAAGAGCCACCA[C/T]GAGATTCACCACAATCCATGGCTGG
Phenotype
MIM: 614423
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ALS2CR11 PubMed Links

Gene Details

Gene
ALS2CR11
Gene Name
amyotrophic lateral sclerosis 2 chromosome region candidate 11
There are no transcripts associated with this gene.

Gene
TMEM237
Gene Name
transmembrane protein 237
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001044385.2 1384 Missense Mutation NP_001037850.1
NM_152388.3 1384 Missense Mutation NP_689601.2

View Full Product Details