Product Details

SNP ID
rs138718796
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:95274727 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACATTCACCCCAGCAGCCAGGGCC[C/T]GGTGGCTGGCCCCTCGAGTTCGTGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PROM2 PubMed Links

Gene Details

Gene
PROM2
Gene Name
prominin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001165977.2 264 Missense Mutation CGG,TGG R48W NP_001159449.1
NM_001165978.2 264 Missense Mutation CGG,TGG R48W NP_001159450.1
NM_001321070.1 264 UTR 5 NP_001307999.1
NM_144707.3 264 Missense Mutation CGG,TGG R48W NP_653308.2
XM_006712301.2 264 Intron XP_006712364.1
XM_011510672.2 264 Missense Mutation CGG,TGG R48W XP_011508974.1

View Full Product Details