Product Details

SNP ID
rs139332124
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:63849755 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCCAGAAGAATGAGAAACAGGGC[A/G]CCAGGCAACAGGGTTTGAGTTCCAG
Phenotype
MIM: 191760 MIM: 613580
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC647115 PubMed Links

Gene Details

Gene
LOC647115
Gene Name
translation initiation factor IF-2
There are no transcripts associated with this gene.

Gene
UGP2
Gene Name
UDP-glucose pyrophosphorylase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001521.1 Intron NP_001001521.1
NM_006759.3 Intron NP_006750.3
XM_005264537.2 Intron XP_005264594.1
XM_005264538.1 Intron XP_005264595.1
XM_006712088.1 Intron XP_006712151.1
XM_011533091.1 Intron XP_011531393.1
XM_017004855.1 Intron XP_016860344.1
XM_017004856.1 Intron XP_016860345.1
XM_017004857.1 Intron XP_016860346.1
Gene
WDPCP
Gene Name
WD repeat containing planar cell polarity effector
There are no transcripts associated with this gene.

View Full Product Details