Product Details

SNP ID
rs139578402
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:55637566 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATACAAATACCTGAATTTCTTGGC[A/C]AACTTCTAATCCTAGGGCAGTAGGA
Phenotype
MIM: 610316
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PNPT1 PubMed Links

Gene Details

Gene
PNPT1
Gene Name
polyribonucleotide nucleotidyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033109.4 2269 Missense Mutation NP_149100.2
XM_005264629.2 2269 Intron XP_005264686.1
XM_017005172.1 2269 Intron XP_016860661.1

View Full Product Details