Product Details

SNP ID
rs140037014
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:182718141 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATCAGAGACTTGAAAAGGATCATT[A/C]TCTGTTTTCTGATAGTGTATATGGC
Phenotype
MIM: 607987
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DNAJC10 PubMed Links

Gene Details

Gene
DNAJC10
Gene Name
DnaJ heat shock protein family (Hsp40) member C10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271581.1 701 Missense Mutation ATC,CTC I19L NP_001258510.1
NM_018981.2 701 Missense Mutation ATC,CTC I19L NP_061854.1

View Full Product Details