Product Details

SNP ID
rs140193720
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:214414021 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTATTTGAGGACTTAGTATCTTCA[A/G]ATTGATCCCAAACAGTTTAAGAGCG
Phenotype
MIM: 612173
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SPAG16 PubMed Links

Gene Details

Gene
SPAG16
Gene Name
sperm associated antigen 16
There are no transcripts associated with this gene.

Gene
VWC2L
Gene Name
von Willebrand factor C domain containing protein 2-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080500.2 Intron NP_001073969.1

View Full Product Details