Product Details

SNP ID
rs140350293
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:86442171 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACGTGGAGCGCGTCGCCGAGTGG[C/T]CCTGGCTCTCCGGGACCATTCGAGC
Phenotype
MIM: 611512
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KDM3A PubMed Links

Gene Details

Gene
KDM3A
Gene Name
lysine demethylase 3A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146688.1 182 Missense Mutation CCC,TCC P42S NP_001140160.1
NM_018433.5 182 Missense Mutation CCC,TCC P42S NP_060903.2
XM_006712051.3 182 UTR 5 XP_006712114.1
XM_017004493.1 182 Missense Mutation CCC,TCC P42S XP_016859982.1
XM_017004494.1 182 UTR 5 XP_016859983.1

View Full Product Details