Product Details

SNP ID
rs140372256
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:208160859 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCGACTTTGGCATCTGCACCCCCC[A/C]AGTCGTGGTACCTTCTGTAGTCCCC
Phenotype
MIM: 615536 MIM: 123660
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C2orf80 PubMed Links

Gene Details

Gene
C2orf80
Gene Name
chromosome 2 open reading frame 80
There are no transcripts associated with this gene.

Gene
CRYGA
Gene Name
crystallin gamma A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014617.3 487 Missense Mutation NP_055432.2
Gene
LOC100507443
Gene Name
uncharacterized LOC100507443
There are no transcripts associated with this gene.

View Full Product Details