Product Details

SNP ID
rs140640160
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:197470147 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAGCCCAGGTCTTCCTGGCTACCC[A/C]AGAACTTGTACCTTGGATTTTTCAG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
COQ10B PubMed Links

Gene Details

Gene
COQ10B
Gene Name
coenzyme Q10B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320818.1 707 Silent Mutation CCA,CCC P125P NP_001307747.1
NM_001320819.1 707 Silent Mutation CCA,CCC P147P NP_001307748.1
NM_001320820.1 707 Silent Mutation CCA,CCC P132P NP_001307749.1
NM_025147.4 707 Silent Mutation CCA,CCC P175P NP_079423.1

View Full Product Details