Product Details

SNP ID
rs141712845
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:233866829 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTCCTTAATTGCAAGAAAAACCT[C/T]AGATGCAGTCACCTTTTTATACTGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MSL3P1 PubMed Links

Gene Details

Gene
MSL3P1
Gene Name
male-specific lethal 3 homolog (Drosophila) pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details